Assay Details
| Gene Symbol: aco2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153959.1 | NP_001147431.1 | 816 | 59 | ||
| GenBank mRNA | BT063513.1 | - | 777 | 59 | ||
| EU955377.1 | - | 816 | 59 | |||
More Information
Set Membership: |
|

aco2
Genomic Map