Assay Details
| Gene Symbol: LOC100282304 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155216.1 | NP_001148688.1 | 999 | 107 | ||
| GenBank mRNA | EU960427.1 | - | 999 | 107 | ||
| EU962563.1 | - | 980 | 107 | |||
More Information
Set Membership: |
|

LOC100282304
Genomic Map