Assay Details
| Gene Symbol: LOC100282811 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155717.1 | NP_001149189.1 | 827 | 84 | ||
| GenBank mRNA | EU962381.1 | - | 827 | 84 | ||
More Information
Set Membership: |
|

LOC100282811
Genomic Map