Assay Details
| Gene Symbol: LOC100282832 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155738.1 | NP_001149210.1 | 382 | 86 | ||
| GenBank mRNA | EU962457.1 | - | 382 | 86 | ||
More Information
Set Membership: |
|

LOC100282832
Genomic Map