Assay Details
| Gene Symbol: LOC100283675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156576.1 | NP_001150048.1 | 358 | 70 | ||
| GenBank mRNA | BT065286.1 | - | 261 | 70 | ||
| EU965504.1 | - | 358 | 70 | |||
More Information
Set Membership: |
|

LOC100283675
Genomic Map