Assay Details
| Gene Symbol: LOC100283905 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156803.1 | NP_001150275.1 | 803 | 69 | ||
| GenBank mRNA | EU966356.1 | - | 803 | 69 | ||
More Information
Set Membership: |
|

LOC100283905
Genomic Map