Assay Details
Gene Symbol: LOC100283946 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001156844.1 | NP_001150316.1 | 734 | 66 | ||
GenBank mRNA | BT039987.1 | - | 420 | 66 | ||
BT088295.1 | - | 395 | 66 | |||
EU953290.1 | - | 432 | 66 | |||
EU953950.1 | - | 439 | 66 | |||
EU959851.1 | - | 439 | 66 | |||
EU966512.1 | - | 734 | 66 | |||
EU966785.1 | - | 469 | 66 | |||
EU976415.1 | - | 439 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
