Assay Details
| Gene Symbol: LOC100284254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157149.1 | NP_001150621.1 | 714 | 94 | ||
| GenBank mRNA | BT054046.1 | - | 706 | 94 | ||
| EU967670.1 | - | 714 | 94 | |||
More Information
Set Membership: |
|

LOC100284254
Genomic Map