Assay Details
| Gene Symbol: LOC100284394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157289.1 | NP_001150761.1 | 208 | 98 | ||
| GenBank mRNA | BT068309.1 | - | 181 | 98 | ||
| EU968142.1 | - | 208 | 98 | |||
More Information
Set Membership: |
|

LOC100284394
Genomic Map