Assay Details
| Gene Symbol: cl3683_1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157343.1 | NP_001150815.1 | 392 | 82 | ||
| GenBank mRNA | BT063810.1 | - | 371 | 82 | ||
| EU968426.1 | - | 392 | 82 | |||
More Information
Set Membership: |
|

cl3683_1
Genomic Map