Assay Details
| Gene Symbol: LOC100284634 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157529.1 | NP_001151001.1 | 376 | 102 | ||
| GenBank mRNA | EU967486.1 | - | 343 | 102 | ||
| EU969109.1 | - | 376 | 102 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100284634| Gene Symbol: LOC100284634 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157529.1 | NP_001151001.1 | 376 | 102 | ||
| GenBank mRNA | EU967486.1 | - | 343 | 102 | ||
| EU969109.1 | - | 376 | 102 | |||