Assay Details
| Gene Symbol: LOC100285669 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158560.1 | NP_001152032.1 | 839 | 87 | ||
| GenBank mRNA | EU973342.1 | - | 839 | 87 | ||
More Information
Set Membership: |
|

LOC100285669
Genomic Map