Assay Details
| Gene Symbol: LOC100274001 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001148387.1 | NP_001141859.1 | 719 | 97 | ||
| GenBank mRNA | BT033932.1 | - | 719 | 97 | ||
| BT042178.1 | - | 735 | 97 | |||
More Information
Set Membership: |
|

LOC100274001
Genomic Map