Assay Details
| Gene Symbol: LOC100275291 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001149410.1 | NP_001142882.1 | 1237 | 97 | ||
| GenBank mRNA | BT068910.1 | - | 1237 | 97 | ||
More Information
Set Membership: |
|

LOC100275291
Genomic Map