Assay Details
| Gene Symbol: LOC100280970 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153890.1 | NP_001147362.1 | 838 | 106 | ||
| GenBank mRNA | EU954969.1 | - | 838 | 106 | ||
More Information
Set Membership: |
|

LOC100280970
Genomic Map