Assay Details
| Gene Symbol: LOC100281564 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154483.1 | NP_001147955.1 | 798 | 91 | ||
| GenBank mRNA | EU957112.1 | - | 798 | 91 | ||
More Information
Set Membership: |
|

LOC100281564
Genomic Map