Assay Details
| Gene Symbol: LOC100281777 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154697.1 | NP_001148169.1 | 629 | 61 | ||
| GenBank mRNA | BT066905.1 | - | 574 | 61 | ||
| EU957893.1 | - | 629 | 61 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100281777| Gene Symbol: LOC100281777 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154697.1 | NP_001148169.1 | 629 | 61 | ||
| GenBank mRNA | BT066905.1 | - | 574 | 61 | ||
| EU957893.1 | - | 629 | 61 | |||