Assay Details
| Gene Symbol: LOC100281777 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154697.1 | NP_001148169.1 | 1384 | 64 | ||
| GenBank mRNA | EU957893.1 | - | 1384 | 64 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100281777| Gene Symbol: LOC100281777 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154697.1 | NP_001148169.1 | 1384 | 64 | ||
| GenBank mRNA | EU957893.1 | - | 1384 | 64 | ||