Assay Details
| Gene Symbol: LOC100282092 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155005.1 | NP_001148477.1 | 628 | 75 | ||
| GenBank mRNA | EU959540.1 | - | 628 | 75 | ||
| EU970197.1 | - | 501 | 75 | |||
More Information
Set Membership: |
|

LOC100282092
Genomic Map