Assay Details
| Gene Symbol: LOC100282326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155238.1 | NP_001148710.1 | 199 | 62 | ||
| GenBank mRNA | BT069638.1 | - | 186 | 62 | ||
| EU960499.1 | - | 199 | 62 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100282326| Gene Symbol: LOC100282326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155238.1 | NP_001148710.1 | 199 | 62 | ||
| GenBank mRNA | BT069638.1 | - | 186 | 62 | ||
| EU960499.1 | - | 199 | 62 | |||