Assay Details
| Gene Symbol: LOC100282813 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155719.1 | NP_001149191.1 | 926 | 69 | ||
| GenBank mRNA | BT068339.1 | - | 892 | 69 | ||
| EU962386.1 | - | 926 | 69 | |||
More Information
Set Membership: |
|

LOC100282813
Genomic Map