Assay Details
| Gene Symbol: LOC100282854 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155760.1 | NP_001149232.1 | 767 | 91 | ||
| GenBank mRNA | EU962534.1 | - | 767 | 91 | ||
More Information
Set Membership: |
|

LOC100282854
Genomic Map