Assay Details
| Gene Symbol: LOC100283153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156055.1 | NP_001149527.1 | 505 | 64 | ||
| GenBank mRNA | BT064619.1 | - | 436 | 64 | ||
| EU963609.1 | - | 505 | 64 | |||
More Information
Set Membership: |
|

LOC100283153
Genomic Map