Assay Details
| Gene Symbol: LOC100283743 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156642.1 | NP_001150114.1 | 966 | 75 | ||
| GenBank mRNA | BT041934.2 | - | 902 | 75 | ||
| EU965769.1 | - | 966 | 75 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100283743| Gene Symbol: LOC100283743 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156642.1 | NP_001150114.1 | 966 | 75 | ||
| GenBank mRNA | BT041934.2 | - | 902 | 75 | ||
| EU965769.1 | - | 966 | 75 | |||