Assay Details
| Gene Symbol: LOC100284543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157438.1 | NP_001150910.1 | 729 | 80 | ||
| GenBank mRNA | EU968796.1 | - | 729 | 80 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100284543| Gene Symbol: LOC100284543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157438.1 | NP_001150910.1 | 729 | 80 | ||
| GenBank mRNA | EU968796.1 | - | 729 | 80 | ||