Assay Details
Gene Symbol: LOC100284543 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157438.1 | NP_001150910.1 | 815 | 61 | ||
GenBank mRNA | EU968796.1 | - | 815 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
