Assay Details
Gene Symbol: LOC100284685 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157580.1 | NP_001151052.1 | 245 | 83 | ||
GenBank mRNA | EU969312.1 | - | 245 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
