Assay Details
| Gene Symbol: LOC100284798 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157693.1 | NP_001151165.1 | 1287 | 89 | ||
| GenBank mRNA | EU969715.1 | - | 1287 | 89 | ||
More Information
Set Membership: |
|

LOC100284798
Genomic Map