Assay Details
| Gene Symbol: LOC100284854 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157749.1 | NP_001151221.1 | 905 | 72 | ||
| GenBank mRNA | BT066380.1 | - | 1436 | 72 | ||
| BT069368.1 | - | 908 | 72 | |||
| EU969912.1 | - | 905 | 72 | |||
More Information
Set Membership: |
|

LOC100284854
Genomic Map