Assay Details
| Gene Symbol: CNR01 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157851.1 | NP_001151323.1 | 262 | 80 | ||
| GenBank mRNA | EU970260.1 | - | 262 | 80 | ||
| HM008653.1 | - | 195 | 80 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CNR01| Gene Symbol: CNR01 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157851.1 | NP_001151323.1 | 262 | 80 | ||
| GenBank mRNA | EU970260.1 | - | 262 | 80 | ||
| HM008653.1 | - | 195 | 80 | |||