Assay Details
| Gene Symbol: LOC100285409 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158302.1 | NP_001151774.1 | 1371 | 69 | ||
| GenBank mRNA | EU972157.1 | - | 1371 | 69 | ||
More Information
Set Membership: |
|

LOC100285409
Genomic Map