Assay Details
| Gene Symbol: LOC100285649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158540.1 | NP_001152012.1 | 349 | 58 | ||
| GenBank mRNA | EU973266.1 | - | 349 | 58 | ||
More Information
Set Membership: |
|

LOC100285649
Genomic Map