Assay Details
| Gene Symbol: LOC100285719 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158610.1 | NP_001152082.1 | 1020 | 64 | ||
| GenBank mRNA | EU961769.1 | - | 203 | 64 | ||
| EU973582.1 | - | 1020 | 64 | |||
More Information
Set Membership: |
|

LOC100285719
Genomic Map