Assay Details
| Gene Symbol: msh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001147243.1 | NP_001140715.1 | 1451 | 77 | ||
| GenBank mRNA | BT039441.1 | - | 1451 | 77 | ||
| BT040725.1 | - | 1333 | 77 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

msh1| Gene Symbol: msh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001147243.1 | NP_001140715.1 | 1451 | 77 | ||
| GenBank mRNA | BT039441.1 | - | 1451 | 77 | ||
| BT040725.1 | - | 1333 | 77 | |||