Assay Details
| Gene Symbol: fab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001147614.1 | NP_001141086.1 | 629 | 99 | ||
| GenBank mRNA | BT040366.1 | - | 629 | 99 | ||
| BT085766.1 | - | 595 | 99 | |||
| BT088241.1 | - | 509 | 99 | |||
| Gene Symbol: LOC100381348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001174213.1 | NP_001167684.1 | 521 | 99 | ||
| GenBank mRNA | EU961218.1 | - | 492 | 99 | ||
| EU964551.1 | - | 521 | 99 | |||
More Information
Set Membership: |
|

fab1
Genomic Map