Assay Details
Gene Symbol: BE518809 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001151680.1 | NP_001145152.1 | 660 | 72 | ||
GenBank mRNA | EU973337.1 | - | 660 | 72 |
Gene Symbol: LOC100382934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001175620.1 | NP_001169091.1 | 701 | 72 | ||
GenBank mRNA | BT065750.1 | - | 701 | 72 |
