Assay Details
| Gene Symbol: LOC100279630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001152626.1 | NP_001146098.1 | 871 | 116 | ||
| GenBank mRNA | BT054617.1 | - | 871 | 116 | ||
More Information
Set Membership: |
|

LOC100279630
Genomic Map