Assay Details
Gene Symbol: LOC100280500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001153420.1 | NP_001146892.1 | 1506 | 91 | ||
GenBank mRNA | BT055137.1 | - | 1498 | 91 | ||
BT069296.1 | - | 1563 | 91 | |||
EU952136.1 | - | 1506 | 91 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
