Assay Details
| Gene Symbol: LOC100281175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154094.1 | NP_001147566.1 | 785 | 70 | ||
| GenBank mRNA | BT069268.1 | - | 785 | 70 | ||
| EU955823.1 | - | 666 | 70 | |||
More Information
Set Membership: |
|

LOC100281175
Genomic Map