Assay Details
| Gene Symbol: LOC100282349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155261.1 | NP_001148733.1 | 190 | 61 | ||
| GenBank mRNA | EU960564.1 | - | 190 | 61 | ||
More Information
Set Membership: |
|

LOC100282349
Genomic Map