Assay Details
| Gene Symbol: LOC100282595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155503.1 | NP_001148975.1 | 241 | 108 | ||
| GenBank mRNA | EU961578.1 | - | 241 | 108 | ||
| EU965625.1 | - | 242 | 108 | |||
More Information
Set Membership: |
|

LOC100282595
Genomic Map