Assay Details
| Gene Symbol: LOC100283171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156073.1 | NP_001149545.1 | 244 | 90 | ||
| GenBank mRNA | EU963667.1 | - | 244 | 90 | ||
More Information
Set Membership: |
|

LOC100283171
Genomic Map