Assay Details
| Gene Symbol: LOC100283908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156806.1 | NP_001150278.1 | 283 | 64 | ||
| GenBank mRNA | BT042214.1 | - | 265 | 64 | ||
| EU966371.1 | - | 283 | 64 | |||
More Information
Set Membership: |
|

LOC100283908
Genomic Map