Assay Details
| Gene Symbol: LOC100284049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156947.1 | NP_001150419.1 | 1532 | 68 | ||
| GenBank mRNA | EU966907.1 | - | 1532 | 68 | ||
More Information
Set Membership: |
|

LOC100284049
Genomic Map