Assay Details
| Gene Symbol: LOC100284648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157543.1 | NP_001151015.1 | 156 | 109 | ||
| GenBank mRNA | BT085847.1 | - | 272 | 109 | ||
| EU969175.1 | - | 156 | 109 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100284648| Gene Symbol: LOC100284648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157543.1 | NP_001151015.1 | 156 | 109 | ||
| GenBank mRNA | BT085847.1 | - | 272 | 109 | ||
| EU969175.1 | - | 156 | 109 | |||