Assay Details
| Gene Symbol: LOC100285133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158027.1 | NP_001151499.1 | 785 | 84 | ||
| GenBank mRNA | EU970973.1 | - | 785 | 84 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100285133| Gene Symbol: LOC100285133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158027.1 | NP_001151499.1 | 785 | 84 | ||
| GenBank mRNA | EU970973.1 | - | 785 | 84 | ||