Assay Details
| Gene Symbol: LOC100285332 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158224.1 | NP_001151696.1 | 345 | 84 | ||
| GenBank mRNA | EU971875.1 | - | 345 | 84 | ||
More Information
Set Membership: |
|

LOC100285332
Genomic Map