Assay Details
| Gene Symbol: LOC100285438 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158331.1 | NP_001151803.1 | 1285 | 72 | ||
| GenBank mRNA | EU972237.1 | - | 1285 | 72 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100285438| Gene Symbol: LOC100285438 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158331.1 | NP_001151803.1 | 1285 | 72 | ||
| GenBank mRNA | EU972237.1 | - | 1285 | 72 | ||