Assay Details
| Gene Symbol: LOC100285585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158476.1 | NP_001151948.1 | 1047 | 80 | ||
| GenBank mRNA | EU972993.1 | - | 1047 | 80 | ||
More Information
Set Membership: |
|

LOC100285585
Genomic Map