Assay Details
| Gene Symbol: RCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111378.1 | NP_001104848.1 | 305 | 52 | ||
| GenBank mRNA | AB021175.1 | - | 305 | 52 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RCP1| Gene Symbol: RCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111378.1 | NP_001104848.1 | 305 | 52 | ||
| GenBank mRNA | AB021175.1 | - | 305 | 52 | ||
Set Membership: |
|